Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mrpl30
Entrez Gene ID
548902
Gene Name
mitochondrial ribosomal protein L30
Gene Aliases
-
Chromosome Location
Chr.2: 109204025 - 109212945 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016148.2 NP_001016148.1 3 - 4 299 81
XM_012956930.2 XP_012812384.1 4 - 5 410 81
XM_012956931.2 XP_012812385.1 4 - 5 320 81
GenBank mRNA CR761990.2 - 3 - 4 299 81

View Full Product Details