Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
grpel1
Entrez Gene ID
548933
Gene Name
GrpE-like 1, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.1: 20356739 - 20362229 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016179.2 NP_001016179.1 3 - 4 348 85
GenBank mRNA BC091625.1 - 3 - 4 348 85
CR761843.2 - 3 - 4 313 85

View Full Product Details