Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dusp19
Entrez Gene ID
549071
Gene Name
dual specificity phosphatase 19
Gene Aliases
dusp17, skrp1, ts-dsp1
Chromosome Location
Chr.9: 56860619 - 56873147 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016317.2 NP_001016317.1 2 - 3 303 81
GenBank mRNA BC158155.1 - 2 - 3 341 81
CR761465.2 - 2 - 3 303 81

View Full Product Details