Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
qtrt2
Entrez Gene ID
549645
Gene Name
queuine tRNA-ribosyltransferase accessory subunit 2
Gene Aliases
qtrtd1
Chromosome Location
Chr.2: 321382 - 333995 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016891.2 NP_001016891.1 5 - 6 692 93
GenBank mRNA BC135739.1 - 5 - 6 600 93
CR848556.2 - 5 - 6 692 93

View Full Product Details