Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mrpl4
Entrez Gene ID
549976
Gene Name
mitochondrial ribosomal protein L4
Gene Aliases
cgi-28, l4mt
Chromosome Location
Chr.3: 126738937 - 126743417 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017222.2 NP_001017222.1 7 - 8 697 127
XM_012953002.2 XP_012808456.1 7 - 8 773 127
GenBank mRNA CR760332.2 - 7 - 8 697 127

View Full Product Details