Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rtcb
Entrez Gene ID
595066
Gene Name
RNA 2',3'-cyclic phosphate and 5'-OH ligase
Gene Aliases
c22orf28
Chromosome Location
Chr.3: 1666409 - 1673957 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030503.1 NP_001025674.1 1 - 2 137 85
GenBank mRNA BC093459.1 - 1 - 2 137 85

View Full Product Details