Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
csde1
Entrez Gene ID
595092
Gene Name
cold shock domain containing E1
Gene Aliases
d1s155e, unr
Chromosome Location
Chr.2: 66977424 - 67022482 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030529.1 NP_001025700.1 16 - 17 2290 63
XM_012963798.2 XP_012819252.1 19 - 20 2645 63
XM_012963809.2 XP_012819263.1 18 - 19 2516 63
XM_018091303.1 XP_017946792.1 19 - 20 2676 63
XM_018091304.1 XP_017946793.1 18 - 19 2664 63
XM_018091305.1 XP_017946794.1 18 - 19 2633 63
XM_018091306.1 XP_017946795.1 18 - 19 2535 63
XM_018091307.1 XP_017946796.1 17 - 18 2523 63
GenBank mRNA BC095920.1 - 16 - 17 2290 63

View Full Product Details