Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lhx5
Entrez Gene ID
548836
Gene Name
LIM homeobox 5
Gene Aliases
Xlim5, xlim-2a, xlim-5
Chromosome Location
Chr.1: 133684609 - 133706969 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016082.2 NP_001016082.1 2 - 3 692 71
GenBank mRNA BC167665.1 - 2 - 3 934 71
BC170608.1 - 2 - 3 513 71
BC170634.1 - 2 - 3 513 71
CR762213.2 - 2 - 3 692 71

View Full Product Details