Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
manf
Entrez Gene ID
549179
Gene Name
mesencephalic astrocyte-derived neurotrophic factor
Gene Aliases
armet, arp
Chromosome Location
Chr.4: 116916094 - 116919409 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016425.2 NP_001016425.1 2 - 3 241 88
GenBank mRNA BC170885.1 - 2 - 3 241 88
BC170889.1 - 2 - 3 241 88
CR761066.2 - 2 - 3 241 88

View Full Product Details