Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
aurka
Entrez Gene ID
549324
Gene Name
aurora kinase A
Gene Aliases
Aurora-A, Eg2, aik, ark1, aur-a, aura, aurora2, btak, pEg2, stk15, stk6, stk7
Chromosome Location
Chr.Un|NW_016683441: 346725 - 385104 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016570.2 NP_001016570.1 - 102 72
GenBank mRNA BC169141.1 - - 151 72
CR760668.2 - - 102 72

View Full Product Details