Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnali1
Entrez Gene ID
549441
Gene Name
dynein axonemal light intermediate chain 1
Gene Aliases
hp28, p28
Chromosome Location
Chr.2: 58952705 - 58962329 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016687.2 NP_001016687.1 3 - 4 421 76
GenBank mRNA BC135942.1 - 3 - 4 420 76
CR760194.2 - 3 - 4 421 76

View Full Product Details