Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rcn1
Entrez Gene ID
549522
Gene Name
reticulocalbin 1
Gene Aliases
pig20, rcal, rcn
Chromosome Location
Chr.4: 9726530 - 9740393 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016768.2 NP_001016768.1 2 - 3 287 93
XM_012961123.2 XP_012816577.1 2 - 3 322 93
XM_012961124.1 XP_012816578.1 2 - 3 346 93
XM_012961125.2 XP_012816579.1 2 - 3 291 93
GenBank mRNA BC158145.1 - 2 - 3 352 93
CR855743.2 - 2 - 3 287 93

View Full Product Details