Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mrpl13
Entrez Gene ID
549550
Gene Name
mitochondrial ribosomal protein L13
Gene Aliases
-
Chromosome Location
Chr.6: 123264280 - 123296103 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012964729.2 XP_012820183.1 4 - 5 416 67
GenBank mRNA BC157157.1 - 4 - 5 534 67
CR855625.2 - 4 - 5 372 67

View Full Product Details