Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ccnq
Entrez Gene ID
549670
Gene Name
cyclin Q
Gene Aliases
fam58a
Chromosome Location
Chr.8: 3478826 - 3496529 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012968449.2 XP_012823903.1 4 - 5 515 101
XM_012968450.2 XP_012823904.1 4 - 5 525 101
XM_018096265.1 XP_017951754.1 4 - 5 545 101
GenBank mRNA CR848270.2 - 4 - 5 602 101

View Full Product Details