Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
taldo1
Entrez Gene ID
549885
Gene Name
transaldolase 1
Gene Aliases
-
Chromosome Location
Chr.4: 36207979 - 36223827 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017131.2 NP_001017131.1 7 - 8 1156 96
GenBank mRNA CR761089.2 - 7 - 8 1156 96

View Full Product Details