Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tbx2
Entrez Gene ID
619588
Gene Name
T-box 2
Gene Aliases
ET, Xltbx2, tbx2-a, tbx2-b, xtbx2
Chromosome Location
Chr.2: 88143815 - 88160164 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018091310.1 XP_017946799.1 3 - 4 1295 79
GenBank mRNA BC135209.1 - 2 - 3 824 79
DQ124206.1 - 2 - 3 995 79

View Full Product Details