Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
psmd7
Entrez Gene ID
447971
Gene Name
proteasome 26S subunit, non-ATPase 7
Gene Aliases
-
Chromosome Location
Chr.4: 62477652 - 62486239 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037272.1 NP_001032349.1 4 - 5 440 85
GenBank mRNA BC077668.1 - 4 - 5 445 85
BC088002.1 - 4 - 5 440 85
CR848186.2 - 4 - 5 389 85

View Full Product Details