Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hebp2
Entrez Gene ID
548365
Gene Name
heme binding protein 2
Gene Aliases
pp23, soul
Chromosome Location
Chr.5: 23827925 - 23831873 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039739.1 NP_001034828.1 3 - 4 477 109
GenBank mRNA BC089188.1 - 3 - 4 444 109
BC118708.1 - 3 - 4 446 109
BC158372.1 - 3 - 4 449 109
CR760976.2 - 3 - 4 477 109

View Full Product Details