Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hsf3
Entrez Gene ID
733835
Gene Name
heat shock transcription factor 3
Gene Aliases
-
Chromosome Location
Chr.8: 19631221 - 19651120 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045588.1 NP_001039053.1 10 - 11 1247 122
GenBank mRNA CR848515.2 - 10 - 11 1247 122

View Full Product Details