Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
enoph1
Entrez Gene ID
733933
Gene Name
enolase-phosphatase 1
Gene Aliases
-
Chromosome Location
Chr.1: 88641233 - 88674443 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045647.1 NP_001039112.1 2 - 3 451 71
XM_012964336.1 XP_012819790.1 2 - 3 71
XM_012964347.2 XP_012819801.1 2 - 3 71
GenBank mRNA BC118728.1 - 2 - 3 451 71
BC135535.1 - 3 - 4 309 71
CR942427.2 - 2 - 3 395 71

View Full Product Details