Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
enoph1
Entrez Gene ID
733933
Gene Name
enolase-phosphatase 1
Gene Aliases
-
Chromosome Location
Chr.1: 88641233 - 88674443 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045647.1 NP_001039112.1 5 - 6 906 104
XM_012964351.2 XP_012819805.1 4 - 5 585 104
GenBank mRNA BC118728.1 - 5 - 6 906 104
BC135535.1 - 5 - 6 631 104
CR942427.2 - 5 - 6 850 104

View Full Product Details