Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc19a2
Entrez Gene ID
734059
Gene Name
solute carrier family 19 (thiamine transporter), member 2
Gene Aliases
-
Chromosome Location
Chr.5: 125152992 - 125160074 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045736.1 NP_001039201.1 2 - 3 990 96
GenBank mRNA CR942394.2 - 2 - 3 990 96

View Full Product Details