Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nrsn1
Entrez Gene ID
779755
Gene Name
neurensin 1
Gene Aliases
vmp
Chromosome Location
Chr.6: 31312722 - 31328404 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078834.1 NP_001072302.1 2 - 3 230 68
GenBank mRNA BC121354.1 - 2 - 3 262 68

View Full Product Details