Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ajap1
Entrez Gene ID
779816
Gene Name
adherens junctions associated protein 1
Gene Aliases
-
Chromosome Location
Chr.7: 89366581 - 89492859 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078895.1 NP_001072363.1 3 - 4 1342 68
GenBank mRNA BC121464.1 - 3 - 4 1342 68

View Full Product Details