Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ephx3
Entrez Gene ID
779884
Gene Name
epoxide hydrolase 3
Gene Aliases
EH3, abhd9
Chromosome Location
Chr.3: 122374381 - 122385601 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078962.1 NP_001072430.1 4 - 5 726 79
GenBank mRNA BC121674.1 - 4 - 5 726 79

View Full Product Details