Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nme5
Entrez Gene ID
780075
Gene Name
NME/NM23 family member 5
Gene Aliases
NM23-X5
Chromosome Location
Chr.3: 22237217 - 22246227 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079151.1 NP_001072619.1 3 - 4 424 73
GenBank mRNA BC123011.1 - 3 - 4 424 73
CU075422.1 - 3 - 4 464 73

View Full Product Details