Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
acaa1
Entrez Gene ID
780143
Gene Name
acetyl-CoA acyltransferase 1
Gene Aliases
acaa, pthio, thio
Chromosome Location
Chr.Un|NW_016683377: 1131864 - 1152789 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079218.1 NP_001072686.1 10 - 11 1078 111
GenBank mRNA BC124019.1 - 10 - 11 1078 111

View Full Product Details