Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cacng7
Entrez Gene ID
780206
Gene Name
calcium channel, voltage-dependent, gamma subunit 7
Gene Aliases
-
Chromosome Location
Chr.7: 108216412 - 108272535 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079281.1 NP_001072749.1 5 - 6 974 77
GenBank mRNA BC125751.1 - 5 - 6 974 77

View Full Product Details