Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mlc1
Entrez Gene ID
780383
Gene Name
megalencephalic leukoencephalopathy with subcortical cysts 1
Gene Aliases
lvm, mlc, mlc1-3
Chromosome Location
Chr.3: 62407297 - 62435070 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079453.1 NP_001072921.1 2 - 3 271 95
XM_012959324.2 XP_012814778.1 1 - 2 1352 95
XM_018092190.1 XP_017947679.1 2 - 3 286 95
GenBank mRNA BC121460.1 - 2 - 3 271 95

View Full Product Details