Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd5a3
Entrez Gene ID
779994
Gene Name
steroid 5 alpha-reductase 3
Gene Aliases
S5AR 3, SRD5alpha3
Chromosome Location
Chr.1: 31179916 - 31184328 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079071.1 NP_001072539.1 2 - 3 529 65
XM_012965419.2 XP_012820873.1 3 - 4 656 65
XM_012965424.2 XP_012820878.1 3 - 4 625 65
GenBank mRNA BC122039.1 - 2 - 3 529 65
CU025196.1 - 3 - 4 626 65

View Full Product Details