Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
clvs1
Entrez Gene ID
100037913
Gene Name
clavesin 1
Gene Aliases
rlbp1l1
Chromosome Location
Chr.6: 102518385 - 102556916 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097346.1 NP_001090815.1 2 - 3 681 67
GenBank mRNA BC135266.1 - 2 - 3 681 67

View Full Product Details