Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prrt1
Entrez Gene ID
100038083
Gene Name
proline rich transmembrane protein 1
Gene Aliases
ng5
Chromosome Location
Chr.8: 54044122 - 54049335 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097355.1 NP_001090824.1 2 - 3 460 74
GenBank mRNA BC127556.1 - 2 - 3 460 74

View Full Product Details