Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
prrt1
Entrez Gene ID
100038083
Gene Name
proline rich transmembrane protein 1
Gene Aliases
ng5
Chromosome Location
Chr.8: 54044122 - 54049335 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097355.1 NP_001090824.1 3 - 4 657 63
XM_018096087.1 XP_017951576.1 3 - 4 780 63
GenBank mRNA BC127556.1 - 3 - 4 657 63

View Full Product Details