Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmtc2
Entrez Gene ID
100124656
Gene Name
transmembrane and tetratricopeptide repeat containing 2
Gene Aliases
-
Chromosome Location
Chr.3: 43698314 - 43867481 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102686.1 NP_001096156.1 9 - 10 2513 84
XM_018091904.1 XP_017947393.1 8 - 9 1576 84
GenBank mRNA BC136039.1 - 9 - 10 2513 84

View Full Product Details