Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
rnaseh1
Entrez Gene ID
100124876
Gene Name
ribonuclease H1
Gene Aliases
h1rna, rnaseh1-a, rnaseh1-b, rnh1
Chromosome Location
Chr.5: 131230587 - 131241209 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102829.1 NP_001096299.1 6 - 7 760 73
XM_012962617.2 XP_012818071.1 6 - 7 989 73
XM_018093760.1 XP_017949249.1 6 - 7 856 73
GenBank mRNA BC135622.1 - 6 - 7 760 73
CT025221.2 - 6 - 7 675 73

View Full Product Details