Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rmc1
Entrez Gene ID
100124886
Gene Name
regulator of MON1-CCZ1
Gene Aliases
c18orf8, wdr98
Chromosome Location
Chr.2: 94121255 - 94154533 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102838.1 NP_001096308.1 18 - 19 1829 102
GenBank mRNA BC135661.1 - 18 - 19 1829 102

View Full Product Details