Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rilpl1
Entrez Gene ID
100124903
Gene Name
Rab interacting lysosomal protein-like 1
Gene Aliases
-
Chromosome Location
Chr.1: 152392110 - 152419375 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102850.1 NP_001096320.1 2 - 3 637 101
GenBank mRNA BC127552.1 - 2 - 3 637 101
BC135712.1 - 2 - 3 621 101
CR942558.2 - 2 - 3 599 101

View Full Product Details