Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
klhdc2
Entrez Gene ID
100124923
Gene Name
kelch domain containing 2
Gene Aliases
-
Chromosome Location
Chr.8: 72204041 - 72207405 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102867.1 NP_001096337.1 2 - 3 360 95
GenBank mRNA BC135774.1 - 2 - 3 360 95

View Full Product Details