The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102942.1 | NP_001096412.1 | 12 - 13 | 1274 | 61 |
XM_018093762.1 | XP_017949251.1 | 12 - 13 | 1307 | 61 | |
GenBank mRNA | BC136067.1 | - | 12 - 13 | 1274 | 61 |
CU075601.1 | - | 12 - 13 | 1239 | 61 |