Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mmp17
Entrez Gene ID
100125088
Gene Name
matrix metallopeptidase 17
Gene Aliases
-
Chromosome Location
Chr.1: 148186995 - 148246646 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102999.1 NP_001096469.1 8 - 9 1551 107
XM_012966783.2 XP_012822237.1 8 - 9 1647 107
GenBank mRNA BC136206.1 - 8 - 9 1551 107

View Full Product Details