Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nip7
Entrez Gene ID
100125143
Gene Name
NIP7, nucleolar pre-rRNA processing protein
Gene Aliases
-
Chromosome Location
Chr.4: 62239758 - 62244969 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103042.1 NP_001096512.1 2 - 3 160 86
GenBank mRNA BC135468.1 - 2 - 3 160 86

View Full Product Details