Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acer1
Entrez Gene ID
100127603
Gene Name
alkaline ceramidase 1
Gene Aliases
asah3
Chromosome Location
Chr.1: 82996859 - 83030452 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112961.2 NP_001106432.1 3 - 4 334 86
XM_012969733.2 XP_012825187.1 3 - 4 553 86
GenBank mRNA EL802545.1 - 3 - 4 358 86

View Full Product Details