Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
haus1
Entrez Gene ID
100127614
Gene Name
HAUS augmin like complex subunit 1
Gene Aliases
ccdc5
Chromosome Location
Chr.1: 193269339 - 193283398 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112969.1 NP_001106440.1 - 839 91
GenBank mRNA BC153729.1 - - 839 91

View Full Product Details