Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
commd2
Entrez Gene ID
100127739
Gene Name
COMM domain containing 2
Gene Aliases
-
Chromosome Location
Chr.5: 118976450 - 118991507 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113076.1 NP_001106547.1 2 - 3 203 94
XM_012962657.2 XP_012818111.1 2 - 3 223 94
XM_012962658.2 XP_012818112.1 2 - 3 282 94
GenBank mRNA BC154925.1 - 2 - 3 203 94

View Full Product Details