Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
alg3
Entrez Gene ID
100127746
Gene Name
ALG3, alpha-1,3- mannosyltransferase
Gene Aliases
-
Chromosome Location
Chr.5: 110979906 - 110987264 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113083.1 NP_001106554.1 8 - 9 1173 66
XM_012962660.2 XP_012818114.1 8 - 9 1045 66
GenBank mRNA BC154933.1 - 8 - 9 1173 66

View Full Product Details