Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cisd3
Entrez Gene ID
100127760
Gene Name
CDGSH iron sulfur domain 3
Gene Aliases
-
Chromosome Location
Chr.2: 51503740 - 51509334 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113091.1 NP_001106562.1 2 - 3 165 67
XM_012956361.2 XP_012811815.1 2 - 3 277 67
GenBank mRNA BC154953.1 - 2 - 3 165 67

View Full Product Details