Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
arpc3
Entrez Gene ID
100038232
Gene Name
actin related protein 2/3 complex subunit 3
Gene Aliases
arc21, p21-arc
Chromosome Location
Chr.1: 154137240 - 154146903 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113666.1 NP_001107138.1 3 - 4 392 179
GenBank mRNA BC157161.1 - 3 - 4 392 179
BC170646.1 - 3 - 4 384 179
BC170650.1 - 3 - 4 384 179
CT025345.2 - 3 - 4 218 179

View Full Product Details