Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
enkur
Entrez Gene ID
100135370
Gene Name
enkurin, TRPC channel interacting protein
Gene Aliases
enkurin
Chromosome Location
Chr.6: 17407828 - 17418309 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114042.1 NP_001107514.1 2 - 3 264 96
GenBank mRNA BC157678.1 - 2 - 3 264 96
BC171058.1 - 2 - 3 256 96
BC171060.1 - 2 - 3 256 96

View Full Product Details