Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fah
Entrez Gene ID
100135385
Gene Name
fumarylacetoacetate hydrolase (fumarylacetoacetase)
Gene Aliases
-
Chromosome Location
Chr.3: 115560720 - 115576485 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114051.1 NP_001107523.1 5 - 6 525 109
GenBank mRNA BC157696.1 - 5 - 6 525 109

View Full Product Details