Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
klf8
Entrez Gene ID
100144679
Gene Name
Kruppel-like factor 8
Gene Aliases
bklf3, dxs741, znf741
Chromosome Location
Chr.8: 18102577 - 18142317 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123440.1 NP_001116912.1 2 - 3 910 79
XM_012967977.2 XP_012823431.1 2 - 3 1237 79
XM_012967978.2 XP_012823432.1 2 - 3 1251 79
XM_018096145.1 XP_017951634.1 3 - 4 1326 79
GenBank mRNA BC161356.1 - 2 - 3 910 79

View Full Product Details